Disc Tab Punches, Electron Microscopy Sciences
Product Details & Documents
These disc punches are produces discs (tabs) from sheet materials. Comes with adjustable side gauge for centering hole of up to 2" maximum reach.
- Maximum thickness for punching is up to 67-mil (0.17 cm)
- Ticket punching type
Ideal for punching adhesive tabs for specimen mounts or producing circle cover slips from ACLAR® films for growing cells.
: The complete unit is supplied with one punch handle and one punching die.
Specifications
- Description:Disc tab punch
- Hole diameter:½"