Pasteur Pipettes, Disposable, Electron Microscopy Sciences
Good for handling infectious materials, inoculating culture media and routine sampling. Available in either Borosilicate or Flint glass, with an outside body diameter of 7 mm.
- Description:Flint
- Length:4.0"L