Orders LinkContactUsLinkComponent
SS Expandable Capsule, Electron Microscopy Sciences
SS Expandable Capsule, Electron Microscopy Sciences
Expandable Capsule
Catalog # 100490-148
Supplier #:  62314-06
SS Expandable Capsule, Electron Microscopy Sciences
Catalog # 100490-148
SS Expandable Capsule, Electron Microscopy Sciences
SS Expandable Capsule, Electron Microscopy Sciences
Expandable Capsule
Price
$345.67
Log in for availability
Pack of 6
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from stainless steel and comes with a positive snap cap. They are expandable. When closed its diameter by depth dimensions are 40 x 7 mm and can be expanded to 11 mm deep. Perforated top and bottom.
Specifications
  • Description:
    Expandable Capsule