Orders LinkContactUsLinkComponent
CD’s Storage Shell and Envelope, Electron Microscopy Sciences
CD’s Storage Shell and Envelope, Electron Microscopy Sciences
C-Shell™ CD Storage
Catalog # 100489-594
Supplier #:  74006-50
CD’s Storage Shell and Envelope, Electron Microscopy Sciences
Catalog # 100489-594
CD’s Storage Shell and Envelope, Electron Microscopy Sciences
CD’s Storage Shell and Envelope, Electron Microscopy Sciences
C-Shell™ CD Storage
Price
$78.85
Log in for availability
Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
C-Shell™ is designed to provide filling, storage, and protection for compact disks or DVD’s during shipping
Paper Sleeves are economical per sleeve and are far less than plastic jewel case. They are space saving, durable and have clarity.
The C-Shell™ is an ultra slim (only 4.5 mm thick), one-piece polypropylene shell with a living hinge.
Specifications
  • Description:
    C-Shell™ CD Storage