Orders LinkContactUsLinkComponent
Bacti-Cinerator IV, Electron Microscopy Sciences
Bacti-Cinerator IV, Electron Microscopy Sciences
Replacement Bacti-Cinerator IV Heat Element, 115 V
Catalog # 101412-636
Supplier #:  62420-55
Bacti-Cinerator IV, Electron Microscopy Sciences
Catalog # 101412-636
Bacti-Cinerator IV, Electron Microscopy Sciences
Bacti-Cinerator IV, Electron Microscopy Sciences
Replacement Bacti-Cinerator IV Heat Element, 115 V
Price
$615.01
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Baci-Cinerator IV sterilizes by infrared heat (Heater Element contains no asbestos) - organic material is incinerated deep inside the ceramic tunnel. To use, simply insert loop or needle into the tunnel. Complete sterilization takes only 5 to 7 seconds at optimum sterilizing temperature of 1500 °F.
  • Prevents infectious spatter and/or cross contamination
  • No open flame -no hazardous gas
  • No need to wait for the loops or needles to turn red
  • Does not require oxygen so it can be used in anaerobic chambers
  • UL approved
Specifications
  • Description:
    Replacement Bacti-Cinerator IV Heat Element
  • Electrical:
    115 V
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Replacement Switch, Bacti-Cinerator III
    Replacement Switch, Bacti-Cinerator III
    100498-070
    Log in for availability
    Each - $114.02