Orders LinkContactUsLinkComponent
Acrylic Plastic Casting, Electron Microscopy Science
Acrylic Plastic Casting, Electron Microscopy Science
Acrylic Plastic Casting Kit, 16 oz
Catalog # 101381-610
Supplier #:  24210-01
Acrylic Plastic Casting, Electron Microscopy Science
Catalog # 101381-610
Acrylic Plastic Casting, Electron Microscopy Science
Acrylic Plastic Casting, Electron Microscopy Science
Acrylic Plastic Casting Kit, 16 oz
Price
$83.27
Log in for availability
Each (1pt US)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Acrylic plastic casting is a thermosetting acrylic-polymer plastic with excellent clarity
Acrylic Plastic Casting: Thermosetting acrylic-polymer plastic with excellent clarity. Place object in a mold, mix and pour. Hard in an hour and object is firmly embedded in clear, stable plastic. Can be polished with soft cloth to high gloss.
Delivery information: The kit consisits of resin and hardener.
Specifications
  • Description:
    Acrylic Plastic Casting Kit
  • Size:
    16 oz