Orders LinkContactUsLinkComponent
Cryo Transport Box, Electron Microscopy Sciences
Cryo Transport Box, Electron Microscopy Sciences
Cryo Transport Box for 5 Tubes
Catalog # 101412-460
Supplier #:  62004-05
Cryo Transport Box, Electron Microscopy Sciences
Catalog # 101412-460
Cryo Transport Box, Electron Microscopy Sciences
Cryo Transport Box, Electron Microscopy Sciences
Cryo Transport Box for 5 Tubes
Price
$148.65
Log in for availability
Pack of 20
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Cryo vial transport box is made from polystyrene and is ideal for transporting cryo vials.
  • Organizes small amounts of cryo vials
  • Durable construction
  • Holds 1.0 to 2.0 mL cryo vials
Specifications
  • Description:
    Cryo Transport Box for 5 Tubes