Orders LinkContactUsLinkComponent
BsgI
BsgI, 250 units
Catalog # 101229-228
Small Business Enterprise
 :  R0559L
BsgI
Catalog # 101229-228
Small Business Enterprise
CAS Number:  
BsgI
BsgI
BsgI, 250 units
CAS Number:  
Price
$343.00
Log in for availability
Each
Product Details & Documents
An E.coli strain that carries the cloned BsgI gene from Bacillus sphaericus B922.
Specifications
Specifications
  • Description:
    BsgI
  • Size:
    250 units
  • Cat. No.:
    101229-228
  • Enzyme Name:
    BsgI
Specifications