Order Entry
United States
Orders LinkContactUsLinkComponent
PvuII

PvuII

PvuII, 50000 units/ml, 5000 units
Catalog # 101228-828
Small Business Enterprise
Supplier #:  R0151T
PvuII
Catalog # 101228-828
Small Business Enterprise
CAS Number:  
PvuII
PvuII

PvuII, 50000 units/ml, 5000 units

CAS Number:  
Price
$75.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

An E.coli strain that carries the PvuII gene from Proteus vulgaris (ATCC 13315).

Specifications

Specifications

  • Concentration:
    50000 units/ml
  • Description:
    PvuII
  • Size:
    5000 units
  • Cat. No.:
    101228-828
  • Enzyme Name:
    PvuII

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.