Order Entry
United States
Orders LinkContactUsLinkComponent
Pacon® Multi-Program Handwriting Paper

Pacon® Multi-Program Handwriting Paper

White, 1?", 8×10.5", 16 lbs.
Catalog # 500019-656
Supplier #:  PAC2418
Pacon® Multi-Program Handwriting Paper
Catalog # 500019-656
Supplier:  
Pacon® Multi-Program Handwriting Paper
Pacon® Multi-Program Handwriting Paper

White, 1?", 8×10.5", 16 lbs.

Discontinued
Product Details & Documents

Rulings conform to Zaner-Bloser™ and/or D’Nealian™ handwriting programs.

  • Blue-dotted midline of Headlines, red base lines

Bright tablet paper ruled on both sides.

Specifications
  • Ruling:
    1?"
  • Color:
    White
  • Dimensions:
    8×10.5"
  • Weight:
    16 lbs.