Order Entry
United States
Orders LinkContactUsLinkComponent
Rhodanine Method for Copper, Electron Microscopy Sciences

Rhodanine Method for Copper, Electron Microscopy Sciences

Rhodamine Saturated Solution, 100 ml
Catalog # 100504-664
Supplier #:  26602-01
Rhodanine Method for Copper, Electron Microscopy Sciences
Catalog # 100504-664
CAS Number:  
Rhodanine Method for Copper, Electron Microscopy Sciences

Rhodamine Saturated Solution, 100 ml

CAS Number:  
Price
$34.64
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specifications

Specifications

  • Description:
    Rhodamine Saturated Solution
  • Size:
    100 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.