Orders LinkContactUsLinkComponent
Precision Pin Vises, Electron Microscopy Sciences
Precision Pin Vises, Electron Microscopy Sciences
Precision pin vise, 4/Set
Catalog # 100497-562
Supplier #:  62074-10
Precision Pin Vises, Electron Microscopy Sciences
Catalog # 100497-562
Precision Pin Vises, Electron Microscopy Sciences
Precision Pin Vises, Electron Microscopy Sciences
Precision pin vise, 4/Set
Price
$56.18
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These tool steel collets grip firmly when screw chucks are tightened.
  • The chromium-plated handle is hollow which allows for the holding of long wires
The body is stepped, with the small portion knurled allowing for rotation between the fingers. The larger portion is smooth and it easily can be held firmly in lathes and drill presses.
Specifications
  • Description:
    Precision pin vise, 4/Set