Orders LinkContactUsLinkComponent
Techni-Stat® Clean Mat, Electron Microscopy Sciences
Techni-Stat® Clean Mat, Electron Microscopy Sciences
Techni State Clean Mat, 2x4ft, 122 cm (4 ft.)
Catalog # 100497-446
Supplier #:  60617-24
Techni-Stat® Clean Mat, Electron Microscopy Sciences
Catalog # 100497-446
Techni-Stat® Clean Mat, Electron Microscopy Sciences
Techni-Stat® Clean Mat, Electron Microscopy Sciences
Techni State Clean Mat, 2x4ft, 122 cm (4 ft.)
Price
$177.67
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Clean Mat is a one-layer 0.060" thick, solid vinyl tablemat designed to be used in a clean room as well as general laboratory areas, where static control is needed.
It is resistant to heat and most oil, grease, and common solvents. This material provides a safe, non-carbon work surface to dissipate a static charge 6.1 x 108Ω/sq. at a workstation. All cut mats come with one snap and a 10' common point ground. The static dissipate rubber design meets EOS/ESD-54.1 @ 10V. (USA).
Specifications
  • Description:
    Techni State Clean Mat, 2x4ft
  • Length:
    122 cm (4 ft.)