Order Entry
United States
Orders LinkContactUsLinkComponent
Permount™ Mounting Medium, Electron Microscopy Science

Permount™ Mounting Medium, Electron Microscopy Science

Permount Mounting Medium, 500 ml
Catalog # 100496-552
Supplier #:  17986-05
Permount™ Mounting Medium, Electron Microscopy Science
Catalog # 100496-552
CAS Number:  
Permount™ Mounting Medium, Electron Microscopy Science
Permount™ Mounting Medium, Electron Microscopy Science

Permount Mounting Medium, 500 ml

CAS Number:  
Price
$222.66
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A toluene-based synthetic resin mounting medium. The right choice for both rapid mounting and long-term storage of slides. Its low viscosity allows for a thinner mounting layer offering better optical quality and bubble-free preparations.

  • Refractive index near that of a fixed protein which helps to keep images free of distortion
  • Permount™ medium is ideal for mounting coverslips to slides with thick or thin specimens
  • Preserves most biological stains with little or no fading when the slides are stored in darkness
  • Contains an anti-oxidant to prevent the formation of annular rings
  • Its high softening point (155°C/311°F) makes it suitable for microprojection

A toluene-based synthetic resin mounting medium. The right choice for both rapid mounting and long term storage of slides. Low viscosity allows for a thinner mounting layer offering better optical quality and bubble-free preparations.
*Ownership of Fisher Scientific

Certifications: Certificates of analysis are available.

Documents

Specifications

Specifications

  • Description:
    Permount Mounting Medium
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.