Micro Cleaning Solution, Electron Microscopy Sciences
Product Details & Documents
Specially designed for ultrasonic cleaning machines. Cleans glass, most metals, plastics, and rubber.These can work with any concentration.
- Biodegradable
- Contains no phosphates
- Removes radioactive contaminants
Not to be used on zinc, magnesium, or aluminum.
Specifications
- Description:Micro Cleaning Solution
- Size:4×1 gal.
- Packaging:Bottle