Orders LinkContactUsLinkComponent
Standard Tungsten Loop Filaments, Electron Microscopy Sciences
Filament Rebuilt, ISI Instruments, ABT, Topcon
Catalog # 100495-392
Standard Tungsten Loop Filaments, Electron Microscopy Sciences
Catalog # 100495-392
Supplier:  
Standard Tungsten Loop Filaments, Electron Microscopy Sciences
Standard Tungsten Loop Filaments, Electron Microscopy Sciences
Filament Rebuilt, ISI Instruments, ABT, Topcon
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tungsten filments with a minimum tip radius for a coherent beam.
  • Rigid attachment of the filament wire to the post
  • Polished post ends
  • Rebuilt many times
  • Better long-term economy
The base assembly of all of our new filaments are guaranteed to be rebuildable for the lifetime of the electron beam instrument. All of our filaments, new or rebuilt, are inspected for consistent quality and performance before being shipped to you. Return the base and we will rebuild the filament. Filament rebuilding services include: reconditioning of the base, cleaning and/or replacement of necessary parts, installment of new standard loop or special filament tip, pre-centering, vacuum normalizing, and microscopic inspection.
Specifications
  • Description:
    Filament Rebuilt
  • For use with:
    ISI Instruments, ABT, Topcon