Orders LinkContactUsLinkComponent
Disc Tab Punches, Electron Microscopy Sciences
Disc Tab Punches, Electron Microscopy Sciences
Disc tab punch, ⁵⁄₁₆"
Catalog # 100494-408
Supplier #:  77850-08
Disc Tab Punches, Electron Microscopy Sciences
Catalog # 100494-408
Disc Tab Punches, Electron Microscopy Sciences
Disc Tab Punches, Electron Microscopy Sciences
Disc tab punch, ⁵⁄₁₆"
Price
$188.12
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These disc punches are produces discs (tabs) from sheet materials. Comes with adjustable side gauge for centering hole of up to 2" maximum reach.
  • Maximum thickness for punching is up to 67-mil (0.17 cm)
  • Ticket punching type
Ideal for punching adhesive tabs for specimen mounts or producing circle cover slips from ACLAR® films for growing cells.
 : The complete unit is supplied with one punch handle and one punching die.
Specifications
  • Description:
    Disc tab punch
  • Hole diameter:
    ⁵⁄₁₆"