Orders LinkContactUsLinkComponent
Perfect Scale, Electron Microscopy Sciences
Perfect Scale, Electron Microscopy Sciences
Scale set, Perfect
Catalog # 100493-000
Supplier #:  70617-10
Perfect Scale, Electron Microscopy Sciences
Catalog # 100493-000
Perfect Scale, Electron Microscopy Sciences
Perfect Scale, Electron Microscopy Sciences
Scale set, Perfect
Price
$247.39
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Perfect scale is made from Nickel alloy and 3 mm in length with 1/10 mm divisions. The Scale fits the Perfect Handle (70616-HD).
Delivery information: The Perfect Scale comes complete with one scale and a handle.
Specifications
  • Description:
    Scale set, Perfect