Orders LinkContactUsLinkComponent
Light Magnifiers, Electron Microscopy Sciences
Light Magnifiers, Electron Microscopy Sciences
Lighted Magnifier
Catalog # 100491-446
Supplier #:  68123
Light Magnifiers, Electron Microscopy Sciences
Catalog # 100491-446
Light Magnifiers, Electron Microscopy Sciences
Light Magnifiers, Electron Microscopy Sciences
Lighted Magnifier
Price
$74.89
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
3.5X Lighted Magnifier comes with a large square magnifier lens which measures 50 mm/side
A flashlight is attached to the magnifier that illuminates poorly lighted background making it bright and clear for easy observation. A variety of types and magnifications are available.
3.5X Lighted Magnifier comes with a large square magnifier lens which measures 50 mm/side. The lens is removable and the flashlight can be used alone.
7X and 10X Lighted Magnifiers are simple and accurate comparator. High quality measuring scale; detachable flashlight.
15X Lighted Magnifier is a high quality magnifier lens. Very convenient to see details under poor lighting conditions.
15X Lighted Scale Magnifier is a scale magnifier 15X is attached to an illuminating device to improve the visibility. Comes with one metric scale. Battery is not included.
The lens is removable and the flashlight can be used alone.
Specifications
  • Magnification:
    15×
  • Description:
    Lighted Magnifier