Orders LinkContactUsLinkComponent
Shell Vials, Electron Microscopy Sciences
Shell Vials, Electron Microscopy Sciences
Shell vials, 3 Dram, Yes
Catalog # 100489-508
Supplier #:  72631-30
Shell Vials, Electron Microscopy Sciences
Catalog # 100489-508
Shell Vials, Electron Microscopy Sciences
Shell Vials, Electron Microscopy Sciences
Shell vials, 3 Dram, Yes
Price
$140.87
Log in for availability
Pack of 144
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These vials are optically clear with straight-sides. Made from Type1, N-51A borosilicate glass. Fixation, dehydration, and infiltration procedures can be done in the same vial. The larger size vials can be used as an embedding media mixture container or for sample storage.
Delivery information: The vials come with polyethylene open bottom snap-caps, Titeseal®, which are packed separately. Except the 4- dram size vial which comes without a closure. We recommend the use of our cork stoppers.
Specifications
  • Cap included:
    Yes
  • Description:
    Shell vials, 3 Dram