Order Entry
United States
Orders LinkContactUsLinkComponent
Pacon® Multi-Program Handwriting Paper

Pacon® Multi-Program Handwriting Paper

White, ½", 8×10.5", 16 lbs.
Catalog # 500019-658
Supplier:   Janitorial Supplies
Supplier #:  PAC2421
Pacon® Multi-Program Handwriting Paper
Catalog # 500019-658
Pacon® Multi-Program Handwriting Paper
Pacon® Multi-Program Handwriting Paper

White, ½", 8×10.5", 16 lbs.

Price
$10.62
Log in for availability
Pack of 500
Minimum order quantity is 1
Product Details & Documents

Rulings conform to Zaner-Bloser™ and/or D’Nealian™ handwriting programs.

  • Blue-dotted midline of Headlines, red base lines

Bright tablet paper ruled on both sides.

Specifications
  • Ruling:
    ½"
  • Color:
    White
  • Dimensions:
    8×10.5"
  • Weight:
    16 lbs.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.