Modular Blocks, Electron Microscopy Sciences
Solid black anodized aluminum blocks for better heat retention.
Block includes thermometer well for monitoring block temperatures. Close contact of tubes to block walls, provides excellent heat transfer. Block puller included with all models. For use with all our brand modular dri-baths. Single block size: 3.75W×2.9D×2" H (9.5×7.4×5.1 cm).
- Model:2075Q
- Description:Modular block
- No. of wells:6
- Holds:25 mm Test Tubes