Modular Blocks, Electron Microscopy Sciences
Product 13259-107 is similar to 101412-738 but manufacturer and specifications may vary.
Product Details & Documents
Solid black anodized aluminum blocks for better heat retention.
Block includes thermometer well for monitoring block temperatures. Close contact of tubes to block walls, provides excellent heat transfer. Block puller included with all models. For use with all our brand modular dri-baths. Single block size: 3.75W×2.9D×2" H (9.5×7.4×5.1 cm).
Specifications
- Model:2071Q
- Description:Modular block
- No. of wells:24
- Holds:10 mm Dia Test Tubes
Accessories
Product Information
- Catalog #ModelHoldsAvailabilityPrice
- Stainless steel cover for models 2064Q and 2065Q