Orders LinkContactUsLinkComponent
Modular Block for Dri-Bath, Electron Microscopy Sciences
Modular Block for Dri-Bath, Electron Microscopy Sciences
Modular Blocks Test Tube, 18 mm
Catalog # 101412-700
Modular Block for Dri-Bath, Electron Microscopy Sciences
Catalog # 101412-700
Supplier:  
Modular Block for Dri-Bath, Electron Microscopy Sciences
Modular Block for Dri-Bath, Electron Microscopy Sciences
Modular Blocks Test Tube, 18 mm
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Fit all our Modular Block Dri-Baths.
  • Anodized coating resists the harshest environments
  • Versatile
  • Solid anodized aluminum blocks provide maximum heat retention
  • All blocks include thermometer well for easily measuring block temperature
  • All blocks are the same size: Blocks measure: 3" (7.6cm) W x 2" (5cm) H x 4" (10.1cm) D
Specifications
  • Description:
    Modular Blocks Test Tube
  • Size:
    18 mm