Order Entry
United States
Orders LinkContactUsLinkComponent
BssHII

BssHII

BssHII, 5000 units/ml, 500 units
Catalog # 101229-016
Small Business Enterprise
Supplier #:  R0199S
BssHII
Catalog # 101229-016
Small Business Enterprise
CAS Number:  
BssHII
BssHII

BssHII, 5000 units/ml, 500 units

CAS Number:  
Price
$83.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

An E.coli strain that carries the BssHII gene from Bacillus stearothermophilus H3 (N. Welker).

Specifications

Specifications

  • Concentration:
    5000 units/ml
  • Description:
    BssHII
  • Size:
    500 units
  • Cat. No.:
    101229-016
  • Enzyme Name:
    BssHII

Specifications