Orders LinkContactUsLinkComponent
Formaldehyde Spill Response™ Kit, Electron Microscopy Sciences
Formaldehyde Spill Response™ Kit, Electron Microscopy Sciences
Formaldehyde Spill Response Refill
Catalog # 100503-182
Supplier #:  24141-R
Formaldehyde Spill Response™ Kit, Electron Microscopy Sciences
Catalog # 100503-182
Formaldehyde Spill Response™ Kit, Electron Microscopy Sciences
Formaldehyde Spill Response™ Kit, Electron Microscopy Sciences
Formaldehyde Spill Response Refill
Price
$376.90
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This kit features FSC-1™ to aid in reducing exposure to formaldehyde gas in the event of a spill. The formalin is transformed into an easily removable semi solid mass. The resulting mass cools rapidly to minimize formaldehyde off gassing. Kits contains 1 – 11oz bottle of FSC-1™ solidifier, 1 pair of vented goggles, a respirator, an open backed gown to protect clothing, a pair of nitrile gloves, 4 scoops and scrapers, 3 wiper pads and 4 thirty gallon bags.
Specifications
  • Description:
    Formaldehyde Spill Response Refill