Water/Ice Bath Rack, Electron Microscopy Sciences
40 Place Rectangular Mailing Rack for 0.5 ml Tubes
Catalog # 100493-768
Supplier: Electron Microscopy Sciences
Supplier #: 72380-24
Water/Ice Bath Rack, Electron Microscopy Sciences
Catalog # 100493-768
Supplier: Electron Microscopy Sciences
Jump to:
Product Details & Documents
Rust-proof anodized aluminum rack – designed to be used in water/ice baths.
- Removable cover keeps tubes closed while boiling
- Handle for easy removal from water baths
- Will not rust
Specifications
- Description:40 Place Rectangular Mailing Rack for 0.5 ml Tubes