Orders LinkContactUsLinkComponent
DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
Pink
Catalog # 100489-268
Supplier #:  71445-P
DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
Catalog # 100489-268
DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
Pink
Price
$51.80
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from high impact polystyrene. This model is similar to the standard DrainRack™ but can hold up to 100 microscope slides in 50 individual numbered slots.
The rack is autoclavable. Dimensions: 75 x 125 x 25 mm (3 x 5 x 1").
Specifications
  • Color:
    Pink