Order Entry
United States
Orders LinkContactUsLinkComponent
Aurion Blocking Solutions, Electron Microscopy Sciences

Aurion Blocking Solutions, Electron Microscopy Sciences

Blocking Solution (Protein A and G), 30 ml, Bottle
Catalog # 100487-982
Supplier #:  25595
Aurion Blocking Solutions, Electron Microscopy Sciences
Catalog # 100487-982
CAS Number:  
Aurion Blocking Solutions, Electron Microscopy Sciences
Aurion Blocking Solutions, Electron Microscopy Sciences

Blocking Solution (Protein A and G), 30 ml, Bottle

CAS Number:  
Price
$140.79
Log in for availability
Each (30ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Aurion blocking solutions effectively block sticky surfaces by multi-point hydrophobic and charge-based interactions. The blocking solutions are specifically designed to meet the characteristics of each type of secondary conjugate.

  • Prevent immunoreagents from binding non-specifically to specimens with sticky surface properties

The signal-to-noise ratio determines the quality of any detection experiment. This ratio is in principle determined by the characteristics of specimen and detection reagents. With adequate blocking and incubation media, background reactions are minimized whereas specific reactions are not hampered.

Procedures to eliminate background comprise three main steps. They are, to suppress residual aldehyde activity, to saturate multipoint hydrophobic moieties and positive charges with high molecular weight compounds such as those present in the AURION blocking solutions. These steps should be balanced for optimum results.

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    Blocking Solution (Protein A and G)
  • Size:
    30 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.