Orders LinkContactUsLinkComponent
Micro Sieve Set, Electron Microscopy Sciences
Micro Sieve Set, Electron Microscopy Sciences
Micro Sieve Set
Catalog # 100496-824
Supplier #:  50110-00
Micro Sieve Set, Electron Microscopy Sciences
Catalog # 100496-824
Micro Sieve Set, Electron Microscopy Sciences
Micro Sieve Set, Electron Microscopy Sciences
Micro Sieve Set
Price
$482.99
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This micro-sieve set consists of a stack of four interchangeable screen-holding sections, a catch pan, and a friction-fit cover. All the parts of the set are very easy to take apart for changing the mesh insert or for cleaning.
  • Entire unit is moulded of poly-propylene
  • Includes phosphor-bronzed mesh inserts in 25, 35, 45, 60, 80, 120, 170, and 230 mesh
  • Holding sections are 2" I.Ø and 6" height
Specifications
  • Description:
    Micro Sieve Set