Fritted Empty Cartridges for Solid Sample Loading, Bonna Agela Technologies
Plastic Empty Flash Column for 330 g
Catalog # 89214-218
Supplier: Phenomenex
Supplier #: FCH330-H
Fritted Empty Cartridges for Solid Sample Loading, Bonna Agela Technologies
Catalog # 89214-218
Supplier: Phenomenex
Jump to:
Product Details & Documents
These empty flash columns contain one column, one cap and two frit plates.
Specifications
- Description:Plastic Empty Flash Column for 330 g