Bottle, HDPE, Rectangular, Ace Glass
1 L (33.8 oz.), HDPE Rectangular Bottle, 53
71000-190
These rectangular shape wide mouth bottles take less shelf space than standard round bottles.
Sturdy, heavy-wall construction for laboratory use and drug storage. Made of high-density polyethylene (HDPE) with leak-proof screw closures. Well suited for shipping and storing both liquids and dry materials.
- Cap Size:53
- Capacity:1 L (33.8 oz.)
- Description:HDPE Rectangular Bottle