Order Entry
United States
Orders LinkContactUsLinkComponent
Bottle, HDPE, Rectangular, Ace Glass

Bottle, HDPE, Rectangular, Ace Glass

500 ml (16.9 oz.), HDPE Rectangular Bottle, 43
Catalog # 71000-188
Supplier:   Ace Glass
Supplier #:  12431-11
Bottle, HDPE, Rectangular, Ace Glass
Catalog # 71000-188
Supplier:  Ace Glass
Bottle, HDPE, Rectangular, Ace Glass
Bottle, HDPE, Rectangular, Ace Glass

500 ml (16.9 oz.), HDPE Rectangular Bottle, 43

Price
$163.78
New Customer Promotional Price
Log in for availability
Pack of 12 Case of 48
Product Details & Documents

These rectangular shape wide mouth bottles take less shelf space than standard round bottles.

Sturdy, heavy-wall construction for laboratory use and drug storage. Made of high-density polyethylene (HDPE) with leak-proof screw closures. Well suited for shipping and storing both liquids and dry materials.

Specifications
  • Cap Size:
    43
  • Capacity:
    500 ml (16.9 oz.)
  • Description:
    HDPE Rectangular Bottle
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.