PYREX® Separating Funnels, Squibb, Glass, Corning
Funnel Only, 6000 ml, GL45
Catalog # 62890-692
Jump to:
Product Details & Documents
Borosilicate glass funnels feature an unbreakable PTFE valve and drip tip and have a screw thread closure at the top. Supplied with a PTFE valve assembly and GL screw cap with PTFE-faced silicone liner. No stopper.
Replacement funnels do not include a PTFE valve and drip tip or cap.
Ordering information: For replacement valve bottoms or caps, contact your Avantor representative.
Documents
Certificates
Search by Lot NumberSpecifications
- Capacity:6000 ml
- Description:Funnel Only
- Thread finish:GL45