Order Entry
United States
ContactUsLinkComponent
Pure* 200 Pipet Tips
Pure™ 200 Pipette Tips, Molecular BioProducts
Catalog # 53225-692
Supplier:  Thermo Scientific
   Low Manufacturing Impact
undefined
Pure™ 200 Pipette Tips, Molecular BioProducts
Catalog # 53225-692
Supplier:  Thermo Scientific
Supplier Number:  3552
   Low Manufacturing Impact

Specifications

  • Volume:
    0.5 - 200 µl
  • Length:
    5.07 cm (2")
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    ISO14001 (Certificate Registration No. EMS 752603)
  • Cat. No.:
    53225-692
  • Supplier no.:
    3552

Specifications

About this item

These tips feature the MicroPoint* design and a 10 µl reference mark for accurate sample handling and precise delivery.

  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
  • Nonsterile tips are autoclavable

Ideal for a wide variety of applications. Tips fit Pipetman*, Eppendorf*, Finnpipette*, Nichiryo*, Rainin*, and Transferpette* pipettors.

Ordering information: Tips 53225-694 and -692 are packaged 96 per tray, 10 trays per pack. Tips 53225-690 are bulk packaged.
Frequently Bought Together