Order Entry
United States
Orders LinkContactUsLinkComponent
Flip Top Bottles

Flip Top Bottles

237 ml (8 oz.), Natural, 24-410, White PP unlined flip top, HDPE
Catalog # 16151-830
Supplier:   Qorpak
Supplier #:  PLC-03410
Flip Top Bottles
Catalog # 16151-830
Supplier:  Qorpak
Flip Top Bottles
Flip Top Bottles

237 ml (8 oz.), Natural, 24-410, White PP unlined flip top, HDPE

Price
$104.54
New Customer Promotional Price
Log in for availability
Pack of 12 Case of 48
Product Details & Documents

Flip top dispensing bottles are translucent and impact resistant.

  • Ideal for dispensing small amounts of liquid
  • Available in HDPE or LDPE formats

Dispensing bottles offer maximum squeezability.

Documents

Specifications
  • Cap Size:
    24-410
  • Cap type:
    White PP unlined flip top
  • Capacity:
    237 ml (8 oz.)
  • Color:
    Natural
  • Material:
    HDPE
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.