Order Entry
United States
Orders LinkContactUsLinkComponent
 

Buffers, Reference Standards, Color-Coded, Ricca Chemical Company

±0.01, Phosphate, 25 °C, Cubetainer®, 20 L, 7.00, Yellow
Catalog # RC1551-5
Supplier:   Ricca Chemical
 :  R1551000-20F
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Catalog # RC1551-5
Supplier:  Ricca Chemical
CAS Number:  
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company

±0.01, Phosphate, 25 °C, Cubetainer®, 20 L, 7.00, Yellow

CAS Number:  
Price
$373.96
Log in for availability
Each (5US Gal)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • Certified traceable to NIST Standard Reference Materials
  • Lot numbers and expiration dates
Specifications

Specifications

  • Pack type:
    Cubetainer®
  • Color:
    Yellow
  • Size:
    20 L
  • pH buffer type:
    Phosphate
  • Reference temperature:
    25 °C
  • pH:
    7.00
  • pH accuracy:
    ±0.01

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.