Order Entry
United States
Orders LinkContactUsLinkComponent
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical

Buffers, Reference standards, Acidic to Neutral, Ricca Chemical

±0.01, Phthalate, 25 °C, Poly bottle, 4 L, 4.00, Colorless
Catalog # RC15001
Supplier:   Ricca Chemical
Supplier #:  R1500000-4A
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Catalog # RC15001
Supplier:  Ricca Chemical
CAS Number:  
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical

±0.01, Phthalate, 25 °C, Poly bottle, 4 L, 4.00, Colorless

CAS Number:  
Price
$144.64
Log in for availability
Each (1US Gal)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Colorless pH buffer solutions are certified to ±0.01 pH at 25°C

  • Range of packaging is available

Each 500 ml plastic bottle is labeled with the certified pH value, the NIST SRM number and a pH/temperature conversion.

Certifications: Standardized against solutions whose certification is traceable to the National Institute of Standards and Technology. Meet USP requirements.

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Color:
    Colorless
  • Size:
    4 L
  • pH buffer type:
    Phthalate
  • Reference temperature:
    25 °C
  • pH:
    4.00
  • pH accuracy:
    ±0.01

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.