Order Entry
United States
Orders LinkContactUsLinkComponent
 

Buffers, Reference standards, Acidic to Neutral, Ricca Chemical

±0.01, HCl, 25 °C, Poly bottle, 500 ml, 1.07, Colorless
Catalog # RC149016
Supplier:   Ricca Chemical
 :  R1490000-500A
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Catalog # RC149016
Supplier:  Ricca Chemical
CAS Number:  
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical

±0.01, HCl, 25 °C, Poly bottle, 500 ml, 1.07, Colorless

CAS Number:  
Price
$49.60
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Colorless pH buffer solutions are certified to ±0.01 pH at 25°C

  • Range of packaging is available

Each 500 ml plastic bottle is labeled with the certified pH value, the NIST SRM number and a pH/temperature conversion.

 : Standardized against solutions whose certification is traceable to the National Institute of Standards and Technology. Meet USP requirements.

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Color:
    Colorless
  • Size:
    500 ml
  • pH buffer type:
    HCl
  • Reference temperature:
    25 °C
  • pH:
    1.07
  • pH accuracy:
    ±0.01

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.