HPLC Columns, YMC Basic
Analytical, C8, 200 Å, 5 µm, 6.00 mm, 250 mm, Silica
Catalog # 97029-582
Supplier: YMC America,Inc
Supplier #: BA99S05-2506WT
Jump to:
Product Details & Documents
The basic columns are unique reversed-phase column designed for analysis of pharmaceuticals and basic compounds.
- Provides high resolution, and excellent peak shape
- Useful for separation of peptides
The neutral silica support used is prepared by an advanced synthetic process which yields a chromatographic material with proprietary pore size and neutral silanols which minimizes interactions with both basic and acidic compounds.
There are no special surface treatments or exotic endcapping procedures so that the columns provides the same general selectivity seen with conventional C18 and C8 columns without basic compounds tailing.
Specifications
- Column Format:Analytical
- Ø int.:6.00 mm
- Particle Size:5 µm
- Pore size:200 Å
- Length:250 mm
- Packing material:Silica
- Phase:C8
- Cat. No.:97029-582
- Carbon Load:7%
- USP Code:L7
- Endcapped:Yes
- pH Range:2 - 7.5



