Dissolution Sampling Cannula, Quality Lab Accessories
1000 mL DV Series Sampling Probe uses QLA "01" style filters in housing for Distek Baths
Catalog # 97002-360
Supplier: QLA
: CAN01K-DV
Jump to:
Product Details & Documents
QLA manufactures a wide variety of Sampling Cannula and Probes for the Dissolution Industry.
- Manual sampling cannula are manufactured in electroplished 316 Stainless Steel or PEEK materials and come in a variety of lengths
Many industry standard cannula configurations and unique, proprietary QLA configurations are available for most manual and automated sampling applications
The USP defines the sampling point as: "A zone half way between the top of the media and top of the paddle or basket not less than 1 cm from the vessel wall. The sampling position is critical but so too is the type of sampling cannula (also known as a probe) used."
Optional stoppers set the proper sampling position. Manual cannula configurations accommodate all standard tip filters.
Specifications
- Description:1000 mL DV Series Sampling Probe uses QLA "01" style filters in housing for Distek Baths