NMC Lockout Pouches
Deluxe Electrical
Catalog # 94004-318
Supplier: ACCUFORM MANUFACTURING, INC
Supplier #: BLOK7
Jump to:
Product Details & Documents
Make lockout/tagout convenient, easily accessible and portable with lockout/tagout kits
Contents include: (1) red nylon pouch (1) 3.81cm (1.5") lockout hasp (1) 1.91cm (0.75") shackle lock (1) single pole circuit breaker lockout (1) double pole circuit breaker lockout (1) single pole circuit breaker lockout (no hole) for all modules.
(10) lockout tags with fasteners for 94004-312 and 94004-318. (5) lockout tags with fasteners for 94004-314 and 94004-316. (1) electrical switch lockout and (1) plug lockout are for 94004-312. (1) oversized breaker lockout is for 94004-318.
Kits contain a variety of lockout devices and accessories to help maintain safety.
Specifications
- Description:Deluxe Electrical