Duramark® Lock Boxes
Metal lock box, 35.5×26.7×17.8 cm (14×10½×7”)
Catalog # 89170-918
Supplier: Therapak, LLC
Supplier #: 36746
Jump to:
Product Details & Documents
Metal lock boxes are constructed of 24 gauge cold-rolled steel, painted with a powder coat finish, and insulated with foam
Plastic lock boxes are made of high impact polystrene and feature a viewing window on the door. Both boxes feature a hinged door that opens downward for convenient access and use as a tray. Two matching keys are supplied. The metal lock box is also available with a combination lock. Over- or under-door straps allow the boxes to be secured outside of a locked door for easy courier access.
Specifications
- Description:Metal lock box
- Dimensions:35.5×26.7×17.8 cm (14×10½×7”)
Accessories
Product Information
- Catalog #AvailabilityPrice
- Under-Door Strap for Metal or Plastic Box
Product Information
- Catalog #AvailabilityPrice
- Over-Door Strap for Metal or Plastic Box
Product Information
- Catalog #AvailabilityPrice
- Extra Long Over-Door Strap for Metal or Plastic BoxExtra Long Over-Door Strap for Metal or Plastic Box10497-106Log in for availabilityEach - $118.05
Product Information
- Catalog #AvailabilityPrice
- Replacement Key for Metal or Plastic Box
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



