Orders LinkContactUsLinkComponent
Ac-DEVD-CHO
Ac-DEVD-CHO
5 mg
Catalog # 89146-636
Supplier:   Enzo Life Sciences
Supplier #:  260-030-M005
Ac-DEVD-CHO
Catalog # 89146-636
Supplier:  Enzo Life Sciences
CAS Number:  
Ac-DEVD-CHO
Ac-DEVD-CHO
5 mg
CAS Number:  
Price
$352.07
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Caspase-3 inhibitor
Highly specific, potent and reversible inhibitor of caspase-3. Ki<1nM for purified caspase-3. IC50=0.2nM for PARP cleavage in cultured human osteosarcoma cell extracts. In contrast, the caspase-1 (ICE) specific inhibitor YVAD-CHO has a Ki=12µM for caspase-3 and IC50>10µM for PARP cleavage in osteosarcoma cell extracts. The DEVD sequence is based on PARP cleavage site Asp-216 for caspase-3.
Documents
Specifications
Specifications
  • Size:
    5 mg
  • Cat. No.:
    89146-636
  • Protein/Peptide Name:
    AC-DEVD-CHO