Order Entry
United States
Orders LinkContactUsLinkComponent
Ac-DEVD-CHO

Ac-DEVD-CHO

1 mg
Catalog # 89146-634
Supplier:   Enzo Life Sciences
Supplier #:  260-030-M001
Ac-DEVD-CHO
Catalog # 89146-634
Supplier:  Enzo Life Sciences
CAS Number:  
Ac-DEVD-CHO
Ac-DEVD-CHO

1 mg

CAS Number:  
Price
$89.38
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Caspase-3 inhibitor

Highly specific, potent and reversible inhibitor of caspase-3. Ki<1nM for purified caspase-3. IC50=0.2nM for PARP cleavage in cultured human osteosarcoma cell extracts. In contrast, the caspase-1 (ICE) specific inhibitor YVAD-CHO has a Ki=12µM for caspase-3 and IC50>10µM for PARP cleavage in osteosarcoma cell extracts. The DEVD sequence is based on PARP cleavage site Asp-216 for caspase-3.

Documents

Specifications

Specifications

  • Size:
    1 mg
  • Cat. No.:
    89146-634
  • Formulation:
    Lyophilized
  • Reconstitution Instructions:
    Add 0.2ml DMSO per 1mg peptide. Shake well until all the contents are dissolved. This furnishes a 10mM solution of this material.
  • Purity:
    >95% (HPLC)
  • Protein/Peptide Name:
    Ac-DEVD-CHO
  • Peptide Content:
    70-90%.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.