Order Entry
United States
Orders LinkContactUsLinkComponent
Human Recombinant Matriptase 2

Human Recombinant Matriptase 2

125 U
Catalog # 89144-978
Supplier:   Enzo Life Sciences
Supplier #:  ALX-201-752-1
Human Recombinant Matriptase 2
Catalog # 89144-978
Supplier:  Enzo Life Sciences
CAS Number:  
Human Recombinant Matriptase 2
Human Recombinant Matriptase 2

125 U

CAS Number:  
Price
$525.38
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Produced in Drosophila Schneider 2 (S2) cells. Recombinant human matriptase-2 (aa 78-811) is fused at the C-terminus to a V5-His-tag.

  • Trypsin-like serine protease.

Matriptase-2 is a potent negative regulator of hepcidin.

Specifications

Specifications

  • Size:
    125 U
  • Cat. No.:
    89144-978
  • Enzyme type:
    Recombinant
  • Form:
    Liquid
  • Formulation:
    In 50 mM HEPES, pH 9.0, containing 400 mM sodium chloride and 20% glycerol.
  • Enzyme Activity:
    ~2.5 U/µl. One unit is defined at the amount of enzyme that releases 1 nmol of AMC per min. at 37 °C in the presence of 50 µM Boc-Gln-Ala-Arg-AMC.
  • Concentration:
    100 nM
  • Species:
    Human
  • Enzyme Name:
    Matriptase 2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.