NEEDLE 25GA 1/2IN SS PK50
Catalog # 89065-572
Supplier: Loctite
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Corning® Treated Culture Dishes, Corning
Supplier: Corning
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
Heavy Duty Nylon Ties, with 50 lbs. Tensile Strength
Supplier: Triton Products
White Nail Clippers, OR Grade, Sklar
Supplier: Sklar