Orders LinkContactUsLinkComponent
Dissolution Sampling Cannula, Quality Lab Accessories
DV Series Sampling Probe Top for Distek Baths
  89049-232
 :   QLA
 :  CANTOP-DV
Dissolution Sampling Cannula, Quality Lab Accessories
  89049-232
 :  QLA
Dissolution Sampling Cannula, Quality Lab Accessories
Dissolution Sampling Cannula, Quality Lab Accessories
DV Series Sampling Probe Top for Distek Baths
$60.59
Each
QLA manufactures a wide variety of Sampling Cannula and Probes for the Dissolution Industry.
  • Manual sampling cannula are manufactured in electroplished 316 Stainless Steel or PEEK materials and come in a variety of lengths
Many industry standard cannula configurations and unique, proprietary QLA configurations are available for most manual and automated sampling applications
The USP defines the sampling point as: "A zone half way between the top of the media and top of the paddle or basket not less than 1 cm from the vessel wall. The sampling position is critical but so too is the type of sampling cannula (also known as a probe) used."
Optional stoppers set the proper sampling position. Manual cannula configurations accommodate all standard tip filters.
  • Description:
    DV Series Sampling Probe Top for Distek Baths