Orders LinkContactUsLinkComponent
Dissolution Sampling Cannula, Quality Lab Accessories
Dissolution Sampling Cannula, Quality Lab Accessories
Distek Style Housing, Adjustable for 1/8" (3.2 mm) Cannula with PTFE Sleeve Cannula Stopper for VanKel EaseAlign Cover
Catalog # 89039-788
Supplier:   QLA
Supplier #:  CA9STP-DV
Dissolution Sampling Cannula, Quality Lab Accessories
Catalog # 89039-788
Supplier:  QLA
Dissolution Sampling Cannula, Quality Lab Accessories
Dissolution Sampling Cannula, Quality Lab Accessories
Distek Style Housing, Adjustable for 1/8" (3.2 mm) Cannula with PTFE Sleeve Cannula Stopper for VanKel EaseAlign Cover
Price
$43.42
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
QLA manufactures a wide variety of Sampling Cannula and Probes for the Dissolution Industry.
  • Manual sampling cannula are manufactured in electroplished 316 Stainless Steel or PEEK materials and come in a variety of lengths
Many industry standard cannula configurations and unique, proprietary QLA configurations are available for most manual and automated sampling applications
The USP defines the sampling point as: "A zone half way between the top of the media and top of the paddle or basket not less than 1 cm from the vessel wall. The sampling position is critical but so too is the type of sampling cannula (also known as a probe) used."
Optional stoppers set the proper sampling position. Manual cannula configurations accommodate all standard tip filters.
Specifications
  • Description:
    Distek Style Housing, Adjustable for 1/8" (3.2 mm) Cannula with PTFE Sleeve Cannula Stopper for VanKel EaseAlign Cover