Order Entry
United States
Orders LinkContactUsLinkComponent
Microscope Slide Coverslips, Polysciences

Microscope Slide Coverslips, Polysciences

Glass cover slips, rectangular, 30×22 mm, 17 mm
Catalog # 87001-916
Supplier:   Polysciences Inc
Supplier #:  23999-1
Microscope Slide Coverslips, Polysciences
Catalog # 87001-916
Supplier:  Polysciences Inc
Microscope Slide Coverslips, Polysciences
Microscope Slide Coverslips, Polysciences

Glass cover slips, rectangular, 30×22 mm, 17 mm

Price
$118.78
Log in for availability
Pack of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Rectangular glass coverslips made of Borosilicate glass. Ideal coverslip for various methods, including routine and special stains, immunohistochemistry, and in situ hybridization.

Specifications
  • Description:
    Glass cover slips, rectangular
  • L×W:
    30×22 mm
  • Thickness:
    17 mm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.